Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161730.1 | NP_001155202.1 | 1 - 2 | 847 | 64 | |
NM_013683.2 | NP_038711.2 | 1 - 2 | 847 | 64 | ||
GenBank mRNA | AK040629.1 | - | 1 - 2 | 676 | 64 | |
AK040919.1 | - | 1 - 2 | 670 | 64 | ||
AK151481.1 | - | 1 - 2 | 602 | 64 | ||
AK152953.1 | - | 1 - 2 | 547 | 64 | ||
AK153890.1 | - | 1 - 2 | 851 | 64 | ||
AK162731.1 | - | 1 - 2 | 647 | 64 | ||
AK166046.1 | - | 1 - 2 | 848 | 64 | ||
AK167485.1 | - | 1 - 2 | 849 | 64 | ||
AK171363.1 | - | 1 - 2 | 602 | 64 | ||
BC013802.1 | - | 1 - 2 | 258 | 3257566 | 64 | |
BC024897.1 | - | 1 - 2 | 370 | 3257734 | 64 | |
KJ650227.1 | - | 1 - 2 | 523 | 64 | ||
M55637.1 | - | 1 - 2 | 239 | 64 | ||
U60018.1 | - | 1 - 2 | 523 | 64 | ||
U60019.1 | - | 1 - 2 | 523 | 64 | ||
U60021.1 | - | 1 - 2 | 523 | 64 | ||
U60022.1 | - | 1 - 2 | 523 | 64 | ||
X59615.1 | - | 1 - 2 | 82 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |