Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113460.2 | NP_001106931.1 | 15 - 16 | 1654 | 74 | |
NM_001113461.2 | NP_001106932.1 | 14 - 15 | 1588 | 74 | ||
NM_001113464.2 | NP_001106935.1 | 14 - 15 | 1567 | 74 | ||
NM_013689.5 | NP_038717.2 | 14 - 15 | 1547 | 74 | ||
GenBank mRNA | AK139329.1 | - | 14 - 15 | 1563 | 74 | |
AK154457.1 | - | 15 - 16 | 1650 | 74 | ||
AK168452.1 | - | 14 - 15 | 1585 | 74 | ||
AK168460.1 | - | 14 - 15 | 1584 | 74 | ||
AK170588.1 | - | 14 - 15 | 1586 | 74 | ||
BC018230.1 | - | 13 - 14 | 1550 | 74 | ||
BC037071.1 | - | 14 - 15 | 1582 | 5029430 | 74 | |
BC062884.1 | - | 10 - 11 | 1130 | 5697182 | 74 | |
S53716.1 | - | 14 - 15 | 1578 | 74 | ||
X55663.1 | - | 14 - 15 | 1582 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |