Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290549.1 | NP_001277478.1 | 7 - 8 | 1384 | 84 | |
NM_001290551.1 | NP_001277480.1 | 6 - 7 | 1231 | 84 | ||
NM_013690.3 | NP_038718.2 | 7 - 8 | 1384 | 84 | ||
GenBank mRNA | BC050824.1 | - | 6 - 7 | 1225 | 6333576 | 84 |
D13738.1 | - | 7 - 8 | 1386 | 84 | ||
S67051.1 | - | 8 - 9 | 1047 | 84 | ||
X67553.1 | - | 7 - 8 | 1155 | 84 | ||
X71426.1 | - | 8 - 9 | 1118 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |