Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130458.1 | NP_001123930.1 | 6 - 7 | 957 | 74 | |
NM_001130459.1 | NP_001123931.1 | 6 - 7 | 926 | 74 | ||
NM_015749.3 | NP_056564.1 | 6 - 7 | 960 | 74 | ||
GenBank mRNA | AF090686.1 | - | 5 - 6 | 887 | 74 | |
AK133707.1 | - | 6 - 7 | 922 | 74 | ||
AK143761.1 | - | 6 - 7 | 928 | 74 | ||
AK147176.1 | - | 6 - 7 | 931 | 74 | ||
AK161618.1 | - | 6 - 7 | 927 | 74 | ||
AK170401.1 | - | 6 - 7 | 946 | 74 | ||
BC003720.1 | - | 5 - 6 | 833 | 3601531 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |