Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293667.1 | NP_001280596.1 | 26 - 27 | 3365 | 79 | |
NM_001293668.1 | NP_001280597.1 | 26 - 27 | 3365 | 79 | ||
NM_015804.3 | NP_056619.1 | 26 - 27 | 3365 | 79 | ||
NR_121603.1 | 26 - 27 | 3365 | 79 | |||
GenBank mRNA | AF156551.1 | - | 26 - 27 | 3157 | 79 | |
AK147497.1 | - | 26 - 27 | 3367 | 79 | ||
AK173088.1 | - | 26 - 27 | 3518 | 79 | ||
BC036322.1 | - | 11 - 12 | 1426 | 79 | ||
BC040068.1 | - | 5 - 6 | 433 | 79 | ||
BC138715.1 | - | 26 - 27 | 3084 | 8861737 | 79 | |
BC145399.1 | - | 26 - 27 | 3085 | 79 | ||
BC145400.1 | - | 26 - 27 | 3085 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |