Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163336.1 | NP_001156808.1 | 18 - 19 | 2926 | 63 | |
NM_001163337.1 | NP_001156809.1 | 18 - 19 | 2926 | 63 | ||
NM_016745.3 | NP_058025.2 | 18 - 19 | 2926 | 63 | ||
GenBank mRNA | AK089487.1 | - | 18 - 19 | 2926 | 63 | |
AK152999.1 | - | 5 - 6 | 798 | 63 | ||
AK155904.1 | - | 18 - 19 | 2927 | 63 | ||
BC017639.1 | - | 18 - 19 | 2840 | 4924519 | 63 | |
BC026147.1 | - | 18 - 19 | 2872 | 4191458 | 63 | |
U49393.1 | - | 18 - 19 | 2787 | 63 | ||
U49394.1 | - | 18 - 19 | 2787 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |