Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016797.4 | NP_058077.2 | 8 - 9 | 1086 | 131 | |
GenBank mRNA | AB019212.1 | - | 8 - 9 | 756 | 131 | |
AF056323.1 | - | 8 - 9 | 698 | 131 | ||
AK013869.1 | - | 8 - 9 | 740 | 131 | ||
AK082927.1 | - | 8 - 9 | 726 | 131 | ||
AK142742.1 | - | 733 | 131 | |||
AK158923.1 | - | 8 - 9 | 1087 | 131 | ||
AK166822.1 | - | 8 - 9 | 756 | 131 | ||
AK169822.1 | - | 8 - 9 | 759 | 131 | ||
BC132123.1 | - | 7 - 8 | 636 | 40130400 | 131 | |
BC132125.1 | - | 7 - 8 | 636 | 40130402 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |