Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016801.3 | NP_058081.2 | 5 - 6 | 371 | 86 | |
GenBank mRNA | AK144399.1 | - | 2 - 3 | 261 | 86 | |
BC047133.1 | - | 5 - 6 | 360 | 86 | ||
BC057892.1 | - | 5 - 6 | 383 | 86 | ||
BC100446.1 | - | 5 - 6 | 406 | 6595634 | 86 | |
D45208.2 | - | 5 - 6 | 371 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |