Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016804.4 | NP_058084.3 | 6 - 7 | 571 | 93 | |
GenBank mRNA | AB041562.1 | - | 6 - 7 | 563 | 93 | |
AF053550.1 | - | 6 - 7 | 544 | 93 | ||
AK087933.1 | - | 6 - 7 | 559 | 93 | ||
AK168000.1 | - | 6 - 7 | 564 | 93 | ||
AK168021.1 | - | 6 - 7 | 93 | |||
AK168045.1 | - | 6 - 7 | 563 | 93 | ||
AK169048.1 | - | 6 - 7 | 567 | 93 | ||
AK179720.1 | - | 6 - 7 | 366 | 93 | ||
AK208117.1 | - | 3 - 4 | 192 | 93 | ||
AK218920.1 | - | 3 - 4 | 192 | 93 | ||
AK220102.1 | - | 3 - 4 | 192 | 93 | ||
BC006641.1 | - | 6 - 7 | 521 | 3483318 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |