Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016804.4 | NP_058084.3 | 7 - 8 | 610 | 67 | |
GenBank mRNA | AB041562.1 | - | 7 - 8 | 602 | 67 | |
AF053550.1 | - | 7 - 8 | 583 | 67 | ||
AK083055.1 | - | 572 | 67 | |||
AK087933.1 | - | 7 - 8 | 598 | 67 | ||
AK168000.1 | - | 7 - 8 | 603 | 67 | ||
AK168045.1 | - | 7 - 8 | 602 | 67 | ||
AK169048.1 | - | 7 - 8 | 606 | 67 | ||
AK208117.1 | - | 4 - 5 | 231 | 67 | ||
AK211256.1 | - | 4 - 5 | 268 | 67 | ||
AK218920.1 | - | 4 - 5 | 231 | 67 | ||
AK220102.1 | - | 4 - 5 | 231 | 67 | ||
BC006641.1 | - | 7 - 8 | 560 | 3483318 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |