Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016866.2 | NP_058562.1 | 13 - 14 | 1445 | 82 | |
GenBank mRNA | AF099988.1 | - | 13 - 14 | 1421 | 82 | |
AK017292.1 | - | 13 - 14 | 1405 | 82 | ||
AK132016.1 | - | 6 - 7 | 778 | 82 | ||
AK148456.1 | - | 13 - 14 | 1482 | 82 | ||
AK163194.1 | - | 3 - 4 | 160 | 82 | ||
BC051964.1 | - | 13 - 14 | 1732 | 5698326 | 82 | |
BC064443.1 | - | 13 - 14 | 1445 | 6843981 | 82 | |
JN368425.1 | - | 9 - 10 | 1026 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |