Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomm40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109748.1 | NP_001103218.1 | 1 - 2 | 450 | 146 | |
NM_016871.2 | NP_058567.2 | 2 - 3 | 352 | 146 | ||
GenBank mRNA | AF043249.1 | - | 1 - 2 | 278 | 146 | |
AF316403.1 | - | 1 - 2 | 285 | 146 | ||
AF316404.1 | - | 1 - 2 | 285 | 146 | ||
AK150019.1 | - | 1 - 2 | 447 | 146 | ||
AK163759.1 | - | 2 - 3 | 336 | 146 | ||
AK164222.1 | - | 2 - 3 | 351 | 146 | ||
AK164570.1 | - | 2 - 3 | 336 | 146 | ||
AK165173.1 | - | 2 - 3 | 347 | 146 | ||
BC100452.1 | - | 2 - 3 | 340 | 6532792 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |