Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomm40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109748.1 | NP_001103218.1 | 6 - 7 | 937 | 63 | |
NM_016871.2 | NP_058567.2 | 7 - 8 | 839 | 63 | ||
GenBank mRNA | AF043249.1 | - | 6 - 7 | 615 | 63 | |
AF316403.1 | - | 6 - 7 | 772 | 63 | ||
AF316404.1 | - | 6 - 7 | 772 | 63 | ||
AK142575.1 | - | 6 - 7 | 533 | 63 | ||
AK150019.1 | - | 6 - 7 | 934 | 63 | ||
AK163759.1 | - | 7 - 8 | 823 | 63 | ||
AK164570.1 | - | 7 - 8 | 823 | 63 | ||
AK165173.1 | - | 7 - 8 | 834 | 63 | ||
AK170991.1 | - | 7 - 8 | 823 | 63 | ||
AK205085.1 | - | 3 - 4 | 269 | 63 | ||
BC100452.1 | - | 7 - 8 | 827 | 6532792 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |