Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163671.1 | NP_001157143.1 | 5 - 6 | 727 | 83 | |
NM_001163672.1 | NP_001157144.1 | 5 - 6 | 727 | 83 | ||
NM_016961.3 | NP_058657.1 | 5 - 6 | 727 | 83 | ||
NM_207692.2 | NP_997575.2 | 5 - 6 | 727 | 83 | ||
GenBank mRNA | AB005664.1 | - | 4 - 5 | 501 | 83 | |
AF052466.1 | - | 4 - 5 | 463 | 83 | ||
AF052467.1 | - | 4 - 5 | 463 | 83 | ||
AF052468.1 | - | 4 - 5 | 463 | 83 | ||
AF052469.1 | - | 4 - 5 | 463 | 83 | ||
AK031959.1 | - | 5 - 6 | 726 | 83 | ||
AK031974.1 | - | 3 - 4 | 281 | 83 | ||
AK090057.1 | - | 5 - 6 | 671 | 83 | ||
AK145083.1 | - | 5 - 6 | 702 | 83 | ||
BC028341.1 | - | 5 - 6 | 709 | 4192796 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |