Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018758.2 | NP_061228.1 | 6 - 7 | 1264 | 58 | |
GenBank mRNA | AF070975.1 | - | 6 - 7 | 1218 | 58 | |
AK045965.1 | - | 6 - 7 | 1265 | 58 | ||
AK082046.1 | - | 1184 | 58 | |||
AK169462.1 | - | 6 - 7 | 1248 | 58 | ||
AK170182.1 | - | 6 - 7 | 1212 | 58 | ||
BC005423.1 | - | 6 - 7 | 1196 | 3487448 | 58 | |
BC009666.1 | - | 6 - 7 | 1208 | 3490710 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |