Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyro3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290800.1 | NP_001277729.1 | 7 - 8 | 1670 | 72 | |
NM_019392.2 | NP_062265.2 | 7 - 8 | 1206 | 72 | ||
GenBank mRNA | AB000828.1 | - | 7 - 8 | 943 | 72 | |
AK141198.1 | - | 7 - 8 | 977 | 72 | ||
BC066058.1 | - | 7 - 8 | 1206 | 6834015 | 72 | |
BC082325.1 | - | 7 - 8 | 1154 | 30356080 | 72 | |
U05683.1 | - | 7 - 8 | 991 | 72 | ||
U18342.1 | - | 7 - 8 | 1123 | 72 | ||
U18343.1 | - | 7 - 8 | 1666 | 72 | ||
U18933.1 | - | 7 - 8 | 1142 | 72 | ||
X78103.1 | - | 7 - 8 | 1150 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |