Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019488.4 | NP_062361.1 | 8 - 9 | 1195 | 109 | |
GenBank mRNA | AF232061.2 | - | 8 - 9 | 1152 | 109 | |
AJ245936.1 | - | 8 - 9 | 1172 | 109 | ||
AK081806.1 | - | 8 - 9 | 1212 | 109 | ||
AK155792.1 | - | 8 - 9 | 1225 | 109 | ||
AK170319.1 | - | 8 - 9 | 1208 | 109 | ||
AY856043.1 | - | 8 - 9 | 1156 | 109 | ||
AY856044.1 | - | 7 - 8 | 1047 | 109 | ||
AY856046.1 | - | 4 - 5 | 577 | 109 | ||
BC021758.1 | - | 4 - 5 | 521 | 109 | ||
BC090993.1 | - | 8 - 9 | 1195 | 6751763 | 109 | |
Y17802.1 | - | 8 - 9 | 1177 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |