Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uso1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019490.1 | NP_062363.1 | 13 - 14 | 1614 | 75 | |
GenBank mRNA | AF096868.1 | - | 12 - 13 | 1402 | 75 | |
AK144638.1 | - | 13 - 14 | 1605 | 75 | ||
AK156070.1 | - | 13 - 14 | 1607 | 75 | ||
BC016069.1 | - | 13 - 14 | 1614 | 4480992 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |