Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Git2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077359.1 | NP_001070827.1 | 16 - 17 | 1700 | 93 | |
NM_001077360.1 | NP_001070828.1 | 15 - 16 | 1696 | 93 | ||
NM_019834.3 | NP_062808.3 | 16 - 17 | 1783 | 93 | ||
GenBank mRNA | AF148693.1 | - | 16 - 17 | 1657 | 93 | |
AK220323.1 | - | 16 - 17 | 1700 | 93 | ||
BC043062.1 | - | 15 - 16 | 1696 | 5705391 | 93 | |
BC056993.1 | - | 15 - 16 | 1696 | 5705391 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |