Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3cg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146200.1 | NP_001139672.1 | 5 - 6 | 2537 | 87 | |
NM_001146201.1 | NP_001139673.1 | 5 - 6 | 2593 | 87 | ||
NM_020272.2 | NP_064668.2 | 5 - 6 | 2714 | 87 | ||
GenBank mRNA | AF208345.1 | - | 4 - 5 | 2393 | 87 | |
AJ249280.1 | - | 5 - 6 | 2714 | 87 | ||
AK040543.1 | - | 5 - 6 | 2499 | 87 | ||
AK132553.1 | - | 5 - 6 | 2539 | 87 | ||
AK137470.1 | - | 5 - 6 | 2446 | 87 | ||
AK137620.1 | - | 5 - 6 | 2595 | 87 | ||
AK143238.1 | - | 2 - 3 | 218 | 87 | ||
AY831679.1 | - | 1 - 2 | 78 | 87 | ||
BC051246.1 | - | 5 - 6 | 2454 | 6514979 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |