Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301692.1 | NP_001288621.1 | 1 - 2 | 166 | 99 | |
NM_021340.4 | NP_067315.1 | 1 - 2 | 166 | 99 | ||
GenBank mRNA | AF076930.1 | - | 1 - 2 | 86 | 99 | |
AK044450.1 | - | 1 - 2 | 118 | 99 | ||
AK044649.1 | - | 1 - 2 | 117 | 99 | ||
AK149272.1 | - | 1 - 2 | 117 | 99 | ||
BC082611.1 | - | 1 - 2 | 103 | 99 | ||
BC086689.1 | - | 1 - 2 | 93 | 99 | ||
BC119055.1 | - | 1 - 2 | 100 | 8733804 | 99 | |
BC144937.1 | - | 1 - 2 | 100 | 99 | ||
CK622127.1 | - | 1 - 2 | 166 | 99 | ||
CK626179.1 | - | 1 - 2 | 104 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |