Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tollip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023764.3 | NP_076253.1 | 2 - 3 | 276 | 68 | |
GenBank mRNA | AJ242971.1 | - | 2 - 3 | 280 | 68 | |
AK015062.1 | - | 219 | 68 | |||
AK016482.1 | - | 2 - 3 | 277 | 68 | ||
AK045422.1 | - | 2 - 3 | 260 | 68 | ||
AK049263.1 | - | 2 - 3 | 298 | 68 | ||
AK049394.1 | - | 2 - 3 | 222 | 68 | ||
AK078391.1 | - | 286 | 68 | |||
AK161312.1 | - | 2 - 3 | 263 | 68 | ||
AK189903.1 | - | 1 - 2 | 50 | 68 | ||
BC053515.1 | - | 2 - 3 | 263 | 68 | ||
BC062139.1 | - | 2 - 3 | 257 | 6334946 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |