Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500003932.mm.1 |
TC0500000397.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500003932.mm.1 |
NM_030721.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003932.mm.1 |
NM_030721.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003944.mm.1 |
TC0500000397.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500003944.mm.1 |
NM_030721.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003944.mm.1 |
NM_030721.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003938.mm.1 |
TC0500000397.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500003938.mm.1 |
NM_030721.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003938.mm.1 |
NM_030721.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003942.mm.1 |
TC0500000397.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500003942.mm.1 |
NM_030721.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003942.mm.1 |
NM_030721.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003943.mm.1 |
TC0500000397.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500003943.mm.1 |
NM_030721.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003943.mm.1 |
NM_030721.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003941.mm.1 |
TC0500000397.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500003941.mm.1 |
NM_030721.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003941.mm.1 |
NM_030721.2 |
Refseq |