Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032397.2 | NP_115773.2 | 2 - 3 | 555 | 96 | |
GenBank mRNA | AF116525.1 | - | 1 - 2 | 583 | 96 | |
AF357239.1 | - | 2 - 3 | 511 | 96 | ||
AK041033.1 | - | 2 - 3 | 556 | 96 | ||
AK043611.1 | - | 2 - 3 | 525 | 96 | ||
AY258143.1 | - | 1 - 2 | 394 | 96 | ||
BC090622.1 | - | 3 - 4 | 898 | 6832359 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |