Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053116.4 | NP_444346.3 | 2 - 3 | 622 | 108 | |
GenBank mRNA | AK043534.1 | - | 2 - 3 | 562 | 108 | |
AK053805.1 | - | 2 - 3 | 623 | 108 | ||
AK054110.1 | - | 2 - 3 | 620 | 108 | ||
BC115811.1 | - | 2 - 3 | 582 | 40105504 | 108 | |
BC115925.1 | - | 2 - 3 | 582 | 40105502 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |