Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfxn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053196.3 | NP_444426.3 | 8 - 9 | 832 | 76 | |
GenBank mRNA | AF325261.1 | - | 8 - 9 | 790 | 76 | |
AK034875.1 | - | 746 | 76 | |||
AK149277.1 | - | 2664 | 76 | |||
AK150722.1 | - | 8 - 9 | 783 | 76 | ||
AK162407.1 | - | 6 - 7 | 2876 | 76 | ||
BC019808.1 | - | 8 - 9 | 830 | 5043343 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |