Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm9sf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080556.3 | NP_542123.3 | 3 - 4 | 511 | 142 | |
GenBank mRNA | AK010325.1 | - | 3 - 4 | 468 | 142 | |
AK030340.1 | - | 3 - 4 | 441 | 142 | ||
AK048012.1 | - | 3 - 4 | 430 | 142 | ||
AK050338.1 | - | 3 - 4 | 458 | 142 | ||
AK075720.1 | - | 3 - 4 | 511 | 142 | ||
AK088905.1 | - | 3 - 4 | 483 | 142 | ||
BC003862.1 | - | 3 - 4 | 450 | 3488326 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |