Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133184.2 | NP_573447.2 | 3 - 4 | 394 | 86 | |
GenBank mRNA | AF251267.1 | - | 3 - 4 | 394 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mus musculus Probe spans exons |