Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps26a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133672.3 | NP_598433.1 | 1 - 2 | 151 | 101 | |
GenBank mRNA | AK147989.1 | - | 1 - 2 | 92 | 101 | |
AK149859.1 | - | 1 - 2 | 76 | 101 | ||
AK152424.1 | - | 1 - 2 | 95 | 101 | ||
AK159783.1 | - | 1 - 2 | 101 | 101 | ||
AK165853.1 | - | 1 - 2 | 95 | 101 | ||
AK168213.1 | - | 1 - 2 | 101 | |||
AK168537.1 | - | 1 - 2 | 106 | 101 | ||
BB857335.1 | - | 1 - 2 | 168 | 101 | ||
BC007148.1 | - | 1 - 2 | 72 | 3485844 | 101 | |
BC058236.1 | - | 1 - 2 | 78 | 101 | ||
S41204.1 | - | 1 - 2 | 98 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |