Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138652.2 | NP_619593.2 | 6 - 7 | 876 | 73 | |
GenBank mRNA | BC109010.1 | - | 6 - 7 | 874 | 73 | |
BC109011.1 | - | 6 - 7 | 876 | 40048684 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |