Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138652.2 | NP_619593.2 | 8 - 9 | 1262 | 94 | |
GenBank mRNA | BC109010.1 | - | 8 - 9 | 1260 | 94 | |
BC109011.1 | - | 8 - 9 | 1262 | 40048684 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Transporter Mammalian Gene Collection Mus musculus Probe spans exons |