Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145146.2 | NP_660128.2 | 11 - 12 | 1467 | 113 | |
GenBank mRNA | AI255825.1 | - | 4 - 5 | 113 | ||
AJ011080.1 | - | 11 - 12 | 1495 | 113 | ||
BC100597.1 | - | 11 - 12 | 1467 | 6767654 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |