Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pld2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK143930.1 | - | 23 - 24 | 2797 | 78 | |
AK170701.1 | - | 23 - 24 | 2789 | 78 | ||
BC068317.1 | - | 23 - 24 | 2767 | 30094606 | 78 | |
U87557.1 | - | 23 - 24 | 2599 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |