Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009269.2 | NP_033295.2 | 7 - 8 | 709 | 74 | |
GenBank mRNA | AF003823.1 | - | 7 - 8 | 713 | 74 | |
AK028713.1 | - | 7 - 8 | 720 | 74 | ||
AK043064.1 | - | 3 - 4 | 288 | 74 | ||
AK053207.1 | - | 7 - 8 | 714 | 74 | ||
AK079578.1 | - | 7 - 8 | 710 | 74 | ||
AK084391.1 | - | 7 - 8 | 713 | 74 | ||
AK084445.1 | - | 7 - 8 | 706 | 74 | ||
AK089030.1 | - | 7 - 8 | 713 | 74 | ||
AK161140.1 | - | 7 - 8 | 746 | 74 | ||
AK161811.1 | - | 3 - 4 | 288 | 74 | ||
BC046323.1 | - | 7 - 8 | 709 | 6391985 | 74 | |
BC068243.1 | - | 3567 | 74 | |||
X95641.1 | - | 7 - 8 | 708 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |