Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009284.2 | NP_033310.2 | 16 - 17 | 2182 | 60 | |
GenBank mRNA | AF481808.1 | - | 15 - 16 | 1897 | 60 | |
AF481809.1 | - | 15 - 16 | 1897 | 60 | ||
AK149563.1 | - | 16 - 17 | 2142 | 60 | ||
AK163053.1 | - | 16 - 17 | 2183 | 60 | ||
BC029318.1 | - | 3 - 4 | 371 | 60 | ||
BC127054.1 | - | 16 - 17 | 1972 | 40109712 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |