Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tkt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009388.6 | NP_033414.1 | 5 - 6 | 776 | 61 | |
GenBank mRNA | AF195533.1 | - | 4 - 5 | 442 | 61 | |
AK002627.1 | - | 5 - 6 | 729 | 61 | ||
AK012794.1 | - | 5 - 6 | 731 | 61 | ||
AK030446.1 | - | 5 - 6 | 759 | 61 | ||
AK140965.1 | - | 5 - 6 | 760 | 61 | ||
AK144146.1 | - | 5 - 6 | 731 | 61 | ||
AK145677.1 | - | 5 - 6 | 729 | 61 | ||
AK146157.1 | - | 5 - 6 | 729 | 61 | ||
AK150139.1 | - | 5 - 6 | 729 | 61 | ||
AK150769.1 | - | 5 - 6 | 725 | 61 | ||
AK150844.1 | - | 5 - 6 | 725 | 61 | ||
AK150856.1 | - | 5 - 6 | 725 | 61 | ||
AK150901.1 | - | 5 - 6 | 729 | 61 | ||
AK151916.1 | - | 5 - 6 | 724 | 61 | ||
AK151969.1 | - | 5 - 6 | 724 | 61 | ||
AK152245.1 | - | 5 - 6 | 724 | 61 | ||
AK152460.1 | - | 5 - 6 | 745 | 61 | ||
AK159922.1 | - | 5 - 6 | 729 | 61 | ||
AK166763.1 | - | 5 - 6 | 763 | 61 | ||
AK167084.1 | - | 5 - 6 | 763 | 61 | ||
AK214732.1 | - | 1 - 2 | 53 | 61 | ||
BC055336.1 | - | 5 - 6 | 703 | 6413263 | 61 | |
U05809.1 | - | 5 - 6 | 724 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |