Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pold1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011131.3 | NP_035261.3 | 12 - 13 | 1553 | 84 | |
GenBank mRNA | AK088310.1 | - | 12 - 13 | 1544 | 84 | |
AK146837.1 | - | 12 - 13 | 1550 | 84 | ||
AK166592.1 | - | 12 - 13 | 1747 | 84 | ||
AK167569.1 | - | 12 - 13 | 1536 | 84 | ||
AK168967.1 | - | 12 - 13 | 1516 | 84 | ||
AK169040.1 | - | 12 - 13 | 1549 | 84 | ||
BC009128.1 | - | 12 - 13 | 1554 | 3257174 | 84 | |
BC052670.1 | - | 11 - 12 | 1520 | 30058314 | 84 | |
Z21848.1 | - | 12 - 13 | 1522 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |