Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou2af1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011136.2 | NP_035266.1 | 3 - 4 | 285 | 59 | |
GenBank mRNA | AK088587.1 | - | 3 - 4 | 285 | 59 | |
AK135080.1 | - | 3 - 4 | 286 | 59 | ||
BC058611.1 | - | 3 - 4 | 266 | 30077096 | 59 | |
U43788.1 | - | 3 - 4 | 285 | 59 | ||
Z54283.1 | - | 3 - 4 | 288 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |