Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130188.1 | NP_001123660.1 | 4 - 5 | 611 | 80 | |
NM_001130189.1 | NP_001123661.1 | 3 - 4 | 503 | 80 | ||
NM_001130190.1 | NP_001123662.1 | 3 - 4 | 503 | 80 | ||
NM_001130191.1 | NP_001123663.1 | 3 - 4 | 503 | 80 | ||
NM_011360.3 | NP_035490.3 | 3 - 4 | 503 | 80 | ||
GenBank mRNA | AB117975.1 | - | 3 - 4 | 336 | 80 | |
AB200282.1 | - | 3 - 4 | 318 | 80 | ||
AF031919.1 | - | 3 - 4 | 359 | 80 | ||
AF103877.1 | - | 4 - 5 | 417 | 80 | ||
AK076102.1 | - | 3 - 4 | 325 | 80 | ||
AK132210.1 | - | 3 - 4 | 380 | 80 | ||
BC012665.1 | - | 3 - 4 | 409 | 4013227 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |