Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcof1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198984.1 | NP_001185913.1 | 20 - 21 | 3259 | 71 | |
NM_011552.3 | NP_035682.1 | 19 - 20 | 3151 | 71 | ||
GenBank mRNA | AF001794.1 | - | 19 - 20 | 3095 | 71 | |
AK089518.1 | - | 20 - 21 | 3227 | 71 | ||
BC021514.1 | - | 20 - 21 | 3274 | 5351783 | 71 | |
BC046608.1 | - | 18 - 19 | 2961 | 71 | ||
BC052669.1 | - | 20 - 21 | 3231 | 71 | ||
BC057342.1 | - | 20 - 21 | 3211 | 6825735 | 71 | |
BC060105.1 | - | 19 - 20 | 3104 | 71 | ||
U81030.1 | - | 19 - 20 | 3039 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |