Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011580.3 | NP_035710.2 | 16 - 17 | 2717 | 82 | |
GenBank mRNA | AK080686.1 | - | 16 - 17 | 2752 | 82 | |
AK145202.1 | - | 16 - 17 | 2747 | 82 | ||
AK163092.1 | - | 16 - 17 | 2744 | 82 | ||
BC042422.1 | - | 16 - 17 | 2719 | 4018176 | 82 | |
BC050917.1 | - | 16 - 17 | 2717 | 6405710 | 82 | |
M87276.1 | - | 16 - 17 | 2743 | 82 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AK089315.1 | - | Chr. - - [-] on Build | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |