Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thbs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011581.3 | NP_035711.2 | 18 - 19 | 3387 | 68 | |
GenBank mRNA | AK132508.1 | - | 18 - 19 | 3217 | 68 | |
AK142552.1 | - | 18 - 19 | 3359 | 68 | ||
BC031843.1 | - | 7 - 8 | 1172 | 68 | ||
BC053702.1 | - | 18 - 19 | 3189 | 30062369 | 68 | |
L07803.1 | - | 18 - 19 | 3215 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |