Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thbs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011582.3 | NP_035712.1 | 1 - 2 | 239 | 140 | |
GenBank mRNA | AF102887.1 | - | 1 - 2 | 179 | 140 | |
BC139414.1 | - | 1 - 2 | 119 | 8862436 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |