Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277903.1 | NP_001264832.1 | 8 - 9 | 365 | 78 | |
NM_001277904.1 | NP_001264833.1 | 7 - 8 | 329 | 78 | ||
NM_011618.2 | NP_035748.1 | 8 - 9 | 362 | 78 | ||
GenBank mRNA | AF020946.1 | - | 8 - 9 | 349 | 78 | |
AJ131711.1 | - | 7 - 8 | 310 | 78 | ||
AK160226.1 | - | 8 - 9 | 363 | 78 | ||
BC138871.1 | - | 8 - 9 | 342 | 8861893 | 78 | |
BC141142.1 | - | 8 - 9 | 345 | 9055754 | 78 | |
BC145449.1 | - | 7 - 8 | 309 | 9054007 | 78 | |
BC145450.1 | - | 7 - 8 | 309 | 9054008 | 78 | |
BC145451.1 | - | 8 - 9 | 345 | 9054009 | 78 | |
U92883.1 | - | 8 - 9 | 346 | 78 | ||
U92884.1 | - | 7 - 8 | 313 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |