Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277903.1 | NP_001264832.1 | 12 - 13 | 758 | 65 | |
NM_001277904.1 | NP_001264833.1 | 11 - 12 | 722 | 65 | ||
NM_011618.2 | NP_035748.1 | 12 - 13 | 755 | 65 | ||
GenBank mRNA | AF020946.1 | - | 12 - 13 | 742 | 65 | |
AJ131711.1 | - | 11 - 12 | 703 | 65 | ||
AK003317.1 | - | 4 - 5 | 337 | 65 | ||
AK160226.1 | - | 12 - 13 | 756 | 65 | ||
BC138871.1 | - | 12 - 13 | 735 | 8861893 | 65 | |
BC141142.1 | - | 12 - 13 | 738 | 9055754 | 65 | |
BC145449.1 | - | 11 - 12 | 702 | 9054007 | 65 | |
BC145450.1 | - | 11 - 12 | 702 | 9054008 | 65 | |
BC145451.1 | - | 12 - 13 | 738 | 9054009 | 65 | |
U92883.1 | - | 12 - 13 | 739 | 65 | ||
U92884.1 | - | 11 - 12 | 706 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |