Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Top3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011624.2 | NP_035754.1 | 10 - 11 | 1376 | 62 | |
GenBank mRNA | AB013603.1 | - | 9 - 10 | 1189 | 62 | |
AB045324.1 | - | 10 - 11 | 1450 | 62 | ||
AK144709.1 | - | 11 - 12 | 1362 | 62 | ||
AK146475.1 | - | 10 - 11 | 1398 | 62 | ||
AK163796.1 | - | 3 - 4 | 385 | 62 | ||
BC031723.1 | - | 10 - 11 | 1376 | 5003002 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |