Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011629.3 | NP_035759.3 | 3 - 4 | 441 | 122 | |
GenBank mRNA | AK146891.1 | - | 3 - 4 | 454 | 122 | |
AK149374.1 | - | 3 - 4 | 356 | 122 | ||
BC090662.1 | - | 3 - 4 | 417 | 30682075 | 122 | |
BC094580.1 | - | 3 - 4 | 359 | 30602939 | 122 | |
L26957.1 | - | 2 - 3 | 122 | |||
U28265.1 | - | 2 - 3 | 250 | 122 | ||
U30482.1 | - | 2 - 3 | 122 | |||
Y11436.1 | - | 2 - 3 | 889 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Nuclear Receptor Mammalian Gene Collection Mus musculus Probe spans exons |