Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyrobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011662.2 | NP_035792.1 | 1 - 2 | 114 | 70 | |
GenBank mRNA | AF024637.1 | - | 1 - 2 | 63 | 70 | |
AK154480.1 | - | 1 - 2 | 113 | 70 | ||
BC056450.1 | - | 1 - 2 | 114 | 6814478 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons |