Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2111459 |
BC005415.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2111459 |
BY648241.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2111459 |
AK162841.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2111459 |
AK008689.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2111459 |
AK089004.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
809646 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
541054 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
83934 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
324081 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
296904 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
834791 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
401672 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
913644 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
538519 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
64034 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
535303 |
XM_487043.4 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
535303 |
XM_987106.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
270794 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
217022 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
106673 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
304799 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
585038 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
528432 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
102345 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
152752 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
94207 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P146187 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P147675 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2111459 |
BC005415.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2111459 |
BY648241.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2111459 |
AK162841.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2111459 |
AK008689.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2111459 |
AK089004.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P302749 |
BC005415.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P302749 |
BY648241.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P302749 |
AK008689.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P302749 |
AK089004.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P302749 |
NM_011706.2 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27854640 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27854624 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27854619 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27854620 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27854635 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27854632 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27854638 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27854647 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27854622 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27854631 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27854645 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27854643 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27854626 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27854625 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27854636 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27854629 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27854615 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07368107 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P147675 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07368089 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07368075 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P146187 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P302749 |
BC005415.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P302749 |
BY648241.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P302749 |
AK008689.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P302749 |
AK089004.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P302749 |
NM_011706.2 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P120699 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P133492 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P147675 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P146187 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09602651 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09415854 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09484954 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09484946 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09484941 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09415853 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09136425 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09415856 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09484944 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09484953 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854757 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854683 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854783 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854663 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854632 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854686 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854765 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854730 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854693 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854771 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854649 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854766 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854596 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854655 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854718 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854751 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854645 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854719 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854793 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854732 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854778 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854723 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854659 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854777 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854601 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854653 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854629 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854657 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854640 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854710 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854760 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854682 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854680 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854666 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854638 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854668 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854774 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854746 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854715 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854687 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854624 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854664 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854713 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854775 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854721 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854734 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854674 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854750 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854685 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854691 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854769 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854748 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854619 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854647 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854635 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854678 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854643 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854787 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854776 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854631 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854773 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854768 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854690 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854704 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854667 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854780 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854689 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854731 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854741 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854605 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854590 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854672 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854626 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854726 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854782 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854688 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27854788 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854635 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09484953 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09603998 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854674 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854782 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854643 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854620 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854755 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09484954 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854655 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09430758 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854619 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854769 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854766 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09221763 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09720952 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854675 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854622 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09573171 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854752 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09545506 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854775 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854684 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854768 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09739459 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854671 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09484947 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09316856 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854617 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854759 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854636 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09462739 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854672 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854793 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854678 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854601 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854750 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854682 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854679 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09270577 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854762 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854765 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854666 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09739465 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09627795 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854659 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854614 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854773 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854663 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854603 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854780 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854653 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854623 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854625 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854624 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854776 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854791 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854629 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854667 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854651 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09484941 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09316852 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09415855 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854771 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854631 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854787 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854605 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09603999 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854647 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854749 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854751 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854788 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09294159 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854668 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09462741 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854772 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27854649 |
- |
- |