Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011960.2 | NP_036090.2 | 2 - 3 | 557 | 88 | |
GenBank mRNA | AF079557.1 | - | 2 - 3 | 306 | 88 | |
AK036656.1 | - | 2 - 3 | 580 | 88 | ||
AK080306.1 | - | 2 - 3 | 277 | 88 | ||
AK164013.1 | - | 2 - 3 | 544 | 88 | ||
BC050892.1 | - | 2 - 3 | 556 | 88 | ||
BC053037.1 | - | 2 - 3 | 540 | 88 | ||
BC059827.1 | - | 2 - 3 | 566 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |